Order Entry
Austria
ContactUsLinkComponent
Epoxomicin ≥95% (by HPLC)
Epoxomicin ≥95% (by HPLC)
Catalog # ENZOBMLPI1270100
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Epoxomicin ≥95% (by HPLC)
Catalog # ENZOBMLPI1270100
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Seq:
    1286
  • Environmentally Preferable:

Specifications

About this item

 :
 :554.7 g/mol
CAS Number:134381-21-8
Formula:C₂₈H₅₀N₄O₇
MDL Number:MFCD03791061