Order Entry
Austria
ContactUsLinkComponent
BCECF AM ester ≥95% (by HPLC), Ultrapure
BCECF AM ester ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52102
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
BCECF AM ester ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52102
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :808.69 g/mol
Storage Temperature:Freezer
CAS Number:117464-70-7
Formula:C₄₂H₄₀O₂₁
MDL Number:MFCD00036969