Order Entry
Austria
ContactUsLinkComponent
Prostaglandin D₂ ≥98% (by HPLC)
Prostaglandin D₂ ≥98% (by HPLC)
Catalog # ENZOBMLPG0050010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Prostaglandin D₂ ≥98% (by HPLC)
Catalog # ENZOBMLPG0050010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

Specification Test Results

  • Purity:
    ≥98% (HPLC)

Specification Test Results

About this item

 :
 :352.5 g/mol
 :GHS07
 :GHS08
Storage Temperature:Freezer
CAS Number:41598-07-6