
(S)-Mephenytoin ≥98% (by TLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:70989-04-7
:
Formula:C₁₂H₁₄N₂O₂
MDL Number:MFCD00270025
:218.3 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Purity:≥98% (TLC)
Specification Test Results
Frequently Bought Together