Order Entry
Austria
ContactUsLinkComponent
Virodhamine ≥98% (by TLC)
Irritant
Virodhamine ≥98% (by TLC)
Catalog # ENZOBMLFA0360005
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Irritant
Virodhamine ≥98% (by TLC)
Catalog # ENZOBMLFA0360005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Purity:
    ≥98% (TLC)

Specification Test Results

About this item

 :
 :347.53 g/mol
 :GHS07
CAS Number:443129-35-9
Formula:C₂₂H₃₇NO₂
MDL Number:MFCD06654562