Order Entry
Austria
ContactUsLinkComponent
12(S),20-Dihydroxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (DiHETE) ≥99% (by HPLC)
12(S),20-Dihydroxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (DiHETE) ≥99% (by HPLC)
Catalog # ENZOBMLD1220050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
12(S),20-Dihydroxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (DiHETE) ≥99% (by HPLC)
Catalog # ENZOBMLD1220050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Seq:
    1202

Specifications

Specification Test Results

  • Concentration:
    50 µg/ml
  • Formulation:
    Oil dissolved in ethanol
  • Purity:
    ≥99% (HPLC)

Specification Test Results

About this item

 :
 :336.5 g/mol
EINECS:200-578-6
UN:1170
 :GHS02
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
CAS Number:89614-44-8
Formula:C₂₀H₃₂O₄