Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
AM-630
AM-630 ≥98% (by TLC)
AM-630 ≥98% (by TLC)
ENZOBMLCR1070010
:
ENZO LIFE SCIENCES
New Product
:
AM-630 ≥98% (by TLC)
ENZOBMLCR1070010
:
ENZO LIFE SCIENCES
New Product
:
Pk:
10 mg
Seq:
2275
Environmentally Preferable:
Purity:
≥98% (TLC)
:
:
504.4 g/mol
:
3077
:
GHS09
:
Ambient
:
164178-33-0
:
C₂₃H₂₅IN₂O₃
:
9 III