Order Entry
Austria
ContactUsLinkComponent
FLUO 3/AM ≥95% (by HPLC), Ultrapure
FLUO 3/AM ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52004
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
FLUO 3/AM ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52004
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :1129.85 g/mol
Storage Temperature:Freezer
CAS Number:121714-22-5
Formula:C₅₁H₅₀Cl₂N₂O₂₃