N-Arachidonoyl-serotonin ≥98% (by TLC)
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
CAS Number:187947-37-1
:
Formula:C₃₀H₄₂N₂O₂
MDL Number:MFCD02179189
Molecular Weight:462.678 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together