Order Entry
Austria
ContactUsLinkComponent
3,4-Dichloroisocoumarin ≥98% (by elemental analysis)
3,4-Dichloroisocoumarin ≥98% (by elemental analysis)
Catalog # ENZOBMLPI1100010
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
3,4-Dichloroisocoumarin ≥98% (by elemental analysis)
Catalog # ENZOBMLPI1100010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :215.029 g/mol
Storage Temperature:Refrigerator
CAS Number:51050-59-0
Formula:C₉H₄Cl₂O₂
MDL Number:MFCD00036960