Order Entry
Austria
ContactUsLinkComponent
Calphostin C ≥99% (by TLC)
Calphostin C ≥99% (by TLC)
Catalog # ENZOBMLEI1980100
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Calphostin C ≥99% (by TLC)
Catalog # ENZOBMLEI1980100
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Seq:
    1286

Specifications

About this item

 :
 :790.774 g/mol
CAS Number:121263-19-2
Formula:C₄₄H₃₈O₁₄
MDL Number:MFCD00133155