Order Entry
Austria
ContactUsLinkComponent
4-Hydroxyphenylretinamide ≥97% (by TLC)
Irritant
Health
4-Hydroxyphenylretinamide ≥97% (by TLC)
Catalog # ENZOBMLGR1030005
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Irritant
Health
4-Hydroxyphenylretinamide ≥97% (by TLC)
Catalog # ENZOBMLGR1030005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Source:
    Synthetic
  • Purity:
    ≥97% (TLC)
  • Appearance:
    Orange solid

Specification Test Results

About this item

 :
 :391.56 g/mol
 :GHS07
 :GHS08
CAS Number:65646-68-6
Formula:C₂₆H₃₃NO₂
MDL Number:MFCD00792674