Order Entry
Austria
ContactUsLinkComponent
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥95% (by HPLC), Ultrapure
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52451
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52451
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :389.381 g/mol
Storage Temperature:Freezer
CAS Number:3326-32-7
Formula:C₂₁H₁₁NO₅S
MDL Number:MFCD00005063