Order Entry
Austria
ContactUsLinkComponent
Cytochalasin B ≥98%
Cytochalasin B ≥98%
Catalog # ENZOBMLT1080005
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
undefined
Cytochalasin B ≥98%
Catalog # ENZOBMLT1080005
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 7(S),20(R)-Dihydroxy-16(R)-methyl-10-phenyl-24-oxa[14]cytochalasa-6(12),13(E),21(E)-triene-1,23-dione
 :
 :479.62 g/mol
CAS Number:14930-96-2
Formula:C₂₉H₃₇NO₅
MDL Number:MFCD00077704