Order Entry
Austria
ContactUsLinkComponent
Propidium iodide ≥95% (by HPLC), Ultrapure
Propidium iodide ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52403
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Propidium iodide ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52403
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :668.4 g/mol
Storage Temperature:Freezer
CAS Number:25535-16-4
Formula:C₂₇H₃₄I₂N₄
MDL Number:MFCD00011921