Sodium dodecyl sulphate (SDS)
Catalog # ENZOBMLKI3410012
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:12 mg
- Seq:2288
- Environmentally Preferable:
Specifications
About this item
Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
:
:288.38 g/mol
Storage Temperature:Refrigerator
UN:2926
ADR:4.1,II
:GHS02
:GHS07
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175