Nω-Methyl-L-arginine monoacetate ≥99% (by TLC)
:53308-83-1
:
:C₇H₁₆N₄O₂·CH₃COOH
:MFCD00069311
:248.28 g/mol
:Refrigerator
Frequently Bought Together
Serological pipettes
: Greiner Bio-One
Blender Bags
: Grade Products
VWR® Serological Pipette, Standard Line
: VWR Collection
Screw caps for laboratory/media bottles
: SciLabware
VWR®, Aspirating Pipettes
: VWR Collection
Ethanol denatured 96% ≥96%, GPR RECTAPUR®
: VWR Chemicals
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
VWR® Water Sampling Bottles, with Tamper Evident Cap
: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
: VWR Collection
Sampling scoops, LaboPlast®
: BURKLE
VWR® Aluminium Foil Seals for PCR
: VWR Collection
Detectable scoops, SteriPlast®
: BURKLE