Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cell Freezing Media
BAMBANKER™ Serum Free Cell Freezing Medium
BAMBANKER™ Serum Free Cell Freezing Medium
BAMBANKER™ Serum Free Cell Freezing Medium
Catalog # WAKF302-14686
Supplier:
FUJIFILM WAKO CHEMICALS
CAS Number:
Expand image
BAMBANKER™ Serum Free Cell Freezing Medium
Catalog # WAKF302-14686
Supplier:
FUJIFILM WAKO CHEMICALS
CAS Number:
Specifications
Pk:
100 mL
Description:
Bambanker cell freezing medium
Specifications
About this item
Product Details
Product Details