Order Entry
Austria
ContactUsLinkComponent
ICP-MS Setup Solution: Be, Ce, Fe, In, Li, Mg, Pb, U at 1 µg/L in 1% HNO₃
ICP-MS Setup Solution: Be, Ce, Fe, In, Li, Mg, Pb, U at 1 µg/L in 1% HNO₃
Catalog # PROOLPENXSUSDIL500
CAS Number:  
ICP-MS Setup Solution: Be, Ce, Fe, In, Li, Mg, Pb, U at 1 µg/L in 1% HNO₃
Catalog # PROOLPENXSUSDIL500
CAS Number:  

Specifications

  • Matrix:
    H₂O/tr HNO₃
  • Pk:
    500 mL
  • Description:
    ICP-MS setup solution (8 components)
  • Hazard class description:
    Corrosive
  • UN No.:
    3264
  • Storage temperature:
    Ambient
  • Elements and Concentration:
    1 µg/ml each of Be, Ce, Fe, In, Li, Mg, Pb, U

Specifications

About this item