Order Entry
Austria
ContactUsLinkComponent
Hard Drinking Water UK Low Level Metals
Hard Drinking Water UK Low Level Metals
Catalog # PROOLGC6028
New Product
CAS Number:  
Hard Drinking Water UK Low Level Metals
Catalog # PROOLGC6028
New Product
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Description:
    Hard drinking water UK low level metals
  • Sodium:
    23,47 mg/L
  • Potassium:
    4,91 mg/L
  • Calcium:
    79,6 mg/L
  • Shipping conditions:
    RT
  • Storage temperature:
    5 °C
  • Cadmium:
    0,970 μg/L
  • Manganese:
    4,74 μg/L
  • Zinc:
    58,2 μg/L

Specifications

About this item