Order Entry
Austria
ContactUsLinkComponent
Electrode Filling Solution, 2 M Lithium Chloride Electrolyte
Electrode Filling Solution, 2 M Lithium Chloride Electrolyte
Catalog # PROOALK-LCL-30-5
CAS Number:  
Electrode Filling Solution, 2 M Lithium Chloride Electrolyte
Catalog # PROOALK-LCL-30-5
CAS Number:  

Specifications

  • Pk:
    150 mL
  • Description:
    2 M Lithium chloride electrolyte, electrode filling solution
  • Storage temperature:
    5 °C

Specifications

About this item

  • Concentration: 2 M
  • Shipping temperature: Room temperature
  • Transport restrictions: Air excepted
  • Product format: Single solution