Order Entry
Austria
ContactUsLinkComponent
Conical Tube Adapter
Conical Tube Adapter
Catalog # BECM392268
Supplier:  Beckman Coulter
Conical Tube Adapter
Catalog # BECM392268
Supplier:  Beckman Coulter

Specifications

  • Description:
    Conical tube adapter, PP, Ø 30 mm, 50 ml
  • Pk:
    6

Specifications

About this item

For use in benchtop and microcentrifuge rotors.

  • Colour: Beige