di-Sodium hydrogen phosphate, anhydrous, bioCERTIFIED™ EP, USP
:7558-79-4
:
:Na₂HPO₄
:141.96 g/mol
- Assay (dried basis, USP):98 - 100.5%
- Assay (dried basis, EP):98 - 101.0%
- Appearance of the solution:To pass test
- Loss on drying (USP):5%
- Loss on drying (EP):1.0%
- Inslouble substances:0.4%
- Monosodium Phosphate:2.5%
- Reducing substances:To pass test
- Chloride (Cl):200 ppm
- Sulphate (SO₄):500 ppm
- Arsenic (As):16 ppm
- Iron:20 ppm
- TAMC:1000 CFU/g
- TYMC:100 CFU/g
- Endotoxins:10 IU/g
- Identification:To pass test
- Residual Solvents:As Reported
Frequently Bought Together