Order Entry
Austria
ContactUsLinkComponent
 
Reference Standard, Potassium Tetraoxalate Dihydrate
Catalog # ACSDPH214.L5
Supplier:  CPACHEM
CAS Number:  
Reference Standard, Potassium Tetraoxalate Dihydrate
Catalog # ACSDPH214.L5
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Pk:
    500 mL
  • CAS No.:
    6100-20-5
  • Description:
    Potassium tetraoxalate dihydrate
  • GHS signal:
    Warning
  • Solubility:
    25 g/L
  • Molecular weight:
    254,20 g/mol
  • Storage temperature:
    15...25 °C
  • pH range:
    1,7 (25 °C, 12,6 g/l in H₂O)

Specifications

About this item

Potassium tetraoxalate dihydrate [pH (S) = 1,67₉ (25 °C)] is a certified secondary standard reference material used as a calibration buffer standard for pH instruments or pH electrodes.

  • The product is delivered with a comprehensive, batch-specific certificate of analysis showing the relevant data for the quality management system
  • This buffer standard has a guaranteed shelf life of four years
  • Intended for extremely precise calibration, qualification and monitoring of pH instruments and routine assessment of their performance

The pH value of this Certipur® buffer solution is qualified using differential potentiometry with two platinum hydrogen electrodes against NIST & PTB primary reference materials pH (S) = 1,679 (25 °C). Potassium tetraoxalate dihydrate buffer is a Certified Reference Material (CRM) offered in neat format. Solution of this standard is prepared in-house following instructions in European Pharmacopoeia (Ph. Eur. 2.2.3).