ContactUsLinkComponent
KIMBLE® Black Phenolic, Screw Thread Caps
KIMBLE® Black Phenolic, Screw Thread Caps
Catalog # WHEA73809-15425
Supplier:  DWK Life Sciences
undefined
KIMBLE® Black Phenolic, Screw Thread Caps
Catalog # WHEA73809-15425
Supplier:  DWK Life Sciences
Specifications
  • Cap Size:
    15-425
  • Colour:
    Black
  • Description:
    Black phenolic, screw thread caps, with LDPE cone liner, Cap style: solid top
  • Material:
    Phenolic resin
  • Pk:
    144
Specifications
About this item
Black phenolic cap has a securely mounted LDPE cone-shaped liner which offers a two part seal. The closure forms a seal around the rim and the polyseal cone forms a seal against the inner diameter of the vial opening.
  • Designed for superior torque retention
  • Stress crack resistant
  • Excellent for sample storage and re-sealing
  • Identified in ASTM Specification E982, Class A requirements
  • Continuous knurling