ContactUsLinkComponent
WHEATON® Screw Closures, Polypropylene
WHEATON® Screw Closures, Polypropylene
Catalog # WHEAW015324
Supplier:  DWK Life Sciences
WHEATON® Screw Closures, Polypropylene
Catalog # WHEAW015324
Supplier:  DWK Life Sciences
Specifications
  • Thread:
    13-425
  • Colour:
    White
  • Description:
    Screw closures, solid top, PTFE
  • Material:
    PP
  • Pk:
    1
Specifications
About this item
Use these Polypropylene caps with glass/plastic bottles or vials.
  • Polyvinyl liners can be used with mild acids, alkalis, solvents and alcohols
  • Foamed polyethylene (PE) liner provides chemical resistance for acids, alkalis, solvents and alcohols
PTFE faced foamed PE liner ideal for use with high purity chemicals, strong acids and solvents.