NANOCOLOR® Photometric Standard Test Kits for Detergents
Supplier: MACHEREY-NAGEL
Certificates
About this item
Photometric test kits for the determination of detergents in water samples.
Ready-to-use reagents for up to 500 determinations
- Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
- Low costs per determination
- Easy dilutions for analysis of higher concentrations
The test kits are evaluated in 10 or 50 mm rectangular cuvettes. They are pre-programmed on the NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Sea water analysis: No
Remark: Measuring range on NANOCOLOR VIS II
Hazardous material: Yes
Sludge reagent set: No
Digestion set: No
NanOx Metal: No
NanOx N: No
Specifications
- Model:MANA91834
- Platform:NANOCOLOR® standard tests
- Storage temperature:15...25 °C
- Shelf life (months):36
- Weight:3750 g (Including packaging)
Specifications
Related Information
Frequently Bought Together