Order Entry
Austria
ContactUsLinkComponent
Brady® All Weather Permanent Adhesive Vinyl Labels for M410, M510, and M511 Printers
Brady® All Weather Permanent Adhesive Vinyl Labels for M410, M510, and M511 Printers
Catalog # BRDY177010
Supplier:  Brady
undefined
Brady® All Weather Permanent Adhesive Vinyl Labels for M410, M510, and M511 Printers
Catalog # BRDY177010
Supplier:  Brady

Specifications

  • Colour:
    Black on white
  • Length:
    76,2 mm
  • Material:
    Vinyl
  • Model:
    B-595
  • Pk:
    100
  • Width:
    25,40 mm

Specifications

About this item

Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady handheld label printer.

  • Ideal for labelling pipes, equipment, shelves, and other industrial surfaces
  • Made with high-tack, permanent acrylic adhesive
  • Compatible with Brady printers BMP41, BMP51, BMP53, M410, M510, and M511
  • For use with ribbon series R6000HF

Brady's low-shrink vinyl label material (B-595) is designed to withstand prolonged exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and grime. Made with high-tack, permanent acrylic adhesive for use on rough, highly-textured, dirty and smooth surfaces alike, especially in cold temperatures. These weather-resistant labels are designed to last up to 10 years in an outdoor environment.