Multipurpose containers
Supplier: Greiner Bio-One
Jump to:
About this item
PS, crystal-clear, with snap lid or screw cap.
- Manufactured under aseptically conditions into a Class II Clean Room environment
- For single use only
- Temperature range –20 to +60 °C
Can be used universally for academic and medical purposes.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
15 Options Available Below
Product Details & Documents
PS, crystal-clear, with snap lid or screw cap.
- Manufactured under aseptically conditions into a Class II Clean Room environment
- For single use only
- Temperature range –20 to +60 °C
Can be used universally for academic and medical purposes.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
- CapacityExt.ØHeightTypeColourCap colourVersionSterileAvailabilityPrice
- GREI189170_700With HDPE screw capWith HDPE screw cap7 ml17,00 mm49,00 mmwithout labelaseptic packedLog in for availability1 * 700 ST191,00 €
- GREI189171_700With HDPE screw capWith HDPE screw cap7 ml17,00 mm49,00 mmwithout labelsterileLog in for availability1 * 700 ST128,00 €
- GREI189175_700With HDPE screw capWith HDPE screw cap7 ml17,00 mm49,00 mmwith plain white labelaseptic packedLog in for availability1 * 700 ST118,00 €
- 216-0595With HDPE screw capWith HDPE screw cap30 ml25,03 mm91,04 mmconical bottom, skirted, without labelaseptic packed-Log in for availability1 * 400 ST105,00 €
- GREI201151_400With HDPE screw capWith HDPE screw cap30 ml24,00 mm90,00 mmconical bottom, skirted, with printed labelaseptic packedLog in for availability1 * 400 ST110,00 €
- 216-0596With HDPE screw capWith HDPE screw cap30 ml25,03 mm91,04 mmconcial bottom, skirted, with plain white labelaseptic packed-Log in for availability1 * 400 ST113,00 €
- 212-3164With HDPE screw capWith HDPE screw cap30 ml25,03 mm91,04 mmconical bottom, skirted, without labelsterile+Log in for availability1 * 400 ST112,00 €
- GREI201171_400With HDPE screw capWith HDPE screw cap30 ml24,00 mm90,00 mmwith printed labelsterileLog in for availability1 * 400 ST183,00 €
- 216-0597With HDPE screw capWith HDPE screw cap30 ml25,03 mm91,04 mmconical bottom, skirted, with white plain labelsterile+Log in for availability1 * 400 ST198,00 €
- GREI219102_300With metal screw capWith metal screw cap60 ml40,00 mm60,00 mmwithout labelaseptic packedLog in for availability1 * 300 ST132,00 €
- GREI224170_160With metal screw capWith metal screw cap100 ml48,00 mm78,00 mmwithout labelaseptic packedLog in for availability1 * 160 ST141,00 €
- 216-0600With metal screw capWith metal screw cap150 ml49,00 mm107,00 mmflat bottom, without labelSilverSilveraseptic packed-Log in for availability1 * 120 ST146,00 €
- 216-0601With metal screw capWith metal screw cap150 ml49,00 mm107,00 mmflat bottom, with white plain labelSilveraseptic packed-Log in for availability1 * 120 ST149,00 €
- 216-0602With metal screw capWith metal screw cap250 ml58,00 mm121,00 mmflat bottom, without labelSilveraseptic packed-Log in for availability1 * 48 ST48,20 €
- 216-0603With metal screw capWith metal screw cap250 ml58,00 mm121,00 mmflat bottom, with white plain labelSilveraseptic packed-Log in for availability1 * 48 ST45,60 €