Express Eingabe
Austria
ContactUsLinkComponent
Overshoes Non woven, PP
Overshoes Non woven, PP
Artikelnr. HOPEH10020
Supplier:  Medicom
undefined
Overshoes Non woven, PP
Artikelnr. HOPEH10020
Supplier:  Medicom

Specifications

  • Schuhgröße:
    XL (400×170 mm)
  • Farbe:
    Blau
  • VE:
    1.000
  • Gewicht:
    40 g/m²

Specifications

About this item

These non woven overshoes are made up of PP with Herringbone print sole.

  • Its polypropylene material is particularly used on smooth surfaces in dry environments
  • Its chevron patterns ensure grip on dry floors
Certifications: ISO 9001 manufacturing site.