0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Potassium chloride
Potassium chloride 0.2 M KCl ≥99.5% volumetric solution, Fluka™
Potassium chloride 0.2 M KCl ≥99.5% volumetric solution, Fluka™
34262-1L
:
Honeywell Chemicals
:
Potassium chloride 0.2 M KCl ≥99.5% volumetric solution, Fluka™
34262-1L
:
Honeywell Chemicals
:
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
Factor:
0.998 - 1.002
:
:
74.55 g/mol
:
Ambient
:
231-211-8
:
7447-40-7
:
KCl
:
MFCD00011360