Order Entry
Austria
ContactUsLinkComponent
Ammonium thiocyanate 1 mol/l (1 N) in aqueous solution Reag. Ph. Eur. 1006701
Ammonium thiocyanate 1 mol/l (1 N) in aqueous solution Reag. Ph. Eur. 1006701
Catalog # 87775.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ammonium thiocyanate 1 mol/l (1 N) in aqueous solution Reag. Ph. Eur. 1006701
Catalog # 87775.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :76.12 g/mol
Storage Temperature:Ambient
EINECS:217-175-6
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428