Order Entry
Austria
Orders LinkContactUsLinkComponent
Ammonia solution 20%

Ammonia solution 20%

Glass bottle for solids, 450 g
Catalog # WAKO634-04661
Corrosive
Irritant
Environment
Marketsource
Supplier:   Wako
Ammonia solution 20%
Catalog # WAKO634-04661
Supplier:  Wako
Marketsource
CAS Number:  1336-21-6
Ammonia solution 20%

Glass bottle for solids, 450 g

CAS Number:  1336-21-6
Price
Inquire For Price
Log in for availability
1 * 450 g
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Density:0.92 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    450 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.