BRAND® BIO-CERT® Robotic Tips, PP
About this item
Pipette tips and filter tips from BRAND offer reliable quality for precise and demanding analyses with the pietting robot liquid handling station.
- Raw materials are free from DiHEMDA und Oleamid
- Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged
- No parting agents or demolding aids are used
- BIO-CERT® Liquid handling quality: Free of human DNA, RNase, DNase, pyrogens, PCR-inhibitors and ATP
For use with pipetting robots, tips are required that are matched to the robot's mode of operation in terms of geometry and surface properties. BRAND robotic pipette tips and robotic filter tips meet all requirements for use with the liquid handling station pipetting robot.
16 Options Available Below
Product Details & Documents
Pipette tips and filter tips from BRAND offer reliable quality for precise and demanding analyses with the pietting robot liquid handling station.
- Raw materials are free from DiHEMDA und Oleamid
- Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged
- No parting agents or demolding aids are used
- BIO-CERT® Liquid handling quality: Free of human DNA, RNase, DNase, pyrogens, PCR-inhibitors and ATP
For use with pipetting robots, tips are required that are matched to the robot's mode of operation in terms of geometry and surface properties. BRAND robotic pipette tips and robotic filter tips meet all requirements for use with the liquid handling station pipetting robot.
Product Family Options
1 to 20 µl
Product Information
- Tip styleVolumeSterileFilterPackedAvailabilityPrice
- Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY1 - 20 µl-+1 pack (960 pieces)Log in for availability1 * 960 ST157,00 €
- Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY1 - 20 µl++1 pack (960 pieces)Log in for availability1 * 960 ST186,00 €
1 to 50 µl
Product Information
- Tip styleVolumeSterileFilterPackedAvailabilityPrice
- Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY1 - 50 µl--1 pack (960 pieces)Log in for availability1 * 960 ST89,90 €
- Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY1 - 50 µl+-1 pack (960 pieces)Log in for availability1 * 960 ST102,00 €
10 to 100 µl
Product Information
- Tip styleVolumeSterileFilterPackedAvailabilityPrice
- Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY10 - 100 µl-+1 pack (960 pieces)Log in for availability1 * 960 ST157,00 €
- Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY10 - 100 µl++1 pack (960 pieces)Log in for availability1 * 960 ST186,00 €
10 to 200 µl
Product Information
- Tip styleVolumeSterileFilterPackedAvailabilityPrice
- Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY10 - 200 µl--1 pack (960 pieces)Log in for availability1 * 960 ST72,00 €
- Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY10 - 200 µl+-1 pack (960 pieces)Log in for availability1 * 960 ST95,80 €
- Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY10 - 200 µl-+1 pack (960 pieces)Log in for availability1 * 960 ST157,00 €
- Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY10 - 200 µl++1 pack (960 pieces)Log in for availability1 * 960 ST186,00 €
10 to 300 µl
Product Information
- Tip styleVolumeSterileFilterPackedAvailabilityPrice
- Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY10 - 300 µl--1 pack (960 pieces)Log in for availability1 * 960 ST72,00 €
- Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY10 - 300 µl+-1 pack (960 pieces)Log in for availability1 * 960 ST95,80 €
40 to 1000 µl
Product Information
- Tip styleVolumeSterileFilterPackedAvailabilityPrice
- Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY40 - 1000 µl--1 pack (960 pieces)Log in for availability1 * 960 ST72,00 €
- Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY40 - 1000 µl+-1 pack (960 pieces)Log in for availability1 * 960 ST95,80 €
- Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY40 - 1000 µl-+1 pack (960 pieces)Log in for availability1 * 960 ST157,00 €
- Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY40 - 1000 µl++1 pack (960 pieces)Log in for availability1 * 960 ST186,00 €