Order Entry
Austria
ContactUsLinkComponent
GF AAS Multi element standard XVIII, 16 components in 5% HNO₃
GF AAS Multi element standard XVIII, 16 components in 5% HNO₃
Catalog # CMPL109500.L1
CAS Number:  
GF AAS Multi element standard XVIII, 16 components in 5% HNO₃
Catalog # CMPL109500.L1
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    HDPE bottle
  • Pk:
    100 mL
  • Description:
    GF AAS Multi element standard XVIII (16 components)
  • Elements and Concentration:
    Ag 10 mg/L, Al 100 mg/L, As 100 mg/L, Ba 50 mg/L, Be 5 mg/L, Cd 5 mg/L, Co 50 mg/L, Cr 20 mg/L, Cu 50 mg/L, Fe 20 mg/L, Mn 20 mg/L, Ni 50 mg/L, Pb 100 mg/L, Sb 100 mg/L, Se 100 mg/L, Tl 100 mg/L

Specifications

About this item

16 components: Ag 10 mg/l, Al 100 mg/l, As 100 mg/l, Ba 50 mg/l, Be 5 mg/l, Cd 5 mg/l, Co 50 mg/l, Cr 20 mg/l, Cu 50 mg/l, Fe 20 mg/l, Mn 20 mg/l, Ni 50 mg/l, Pb 100 mg/l, Sb 100 mg/l, Se 100 mg/l, Tl 100 mg/l in 5% HNO₃.

Shelf life upto 36 Months, Store under normal laboratory conditions, at temperatures between 15 °C to 25 °C.

Certifications: ISO 17025/17034/9001