Wavelength Calibration Solution for ICP-OES
Artikelnr. CMPL6610030000.L5
Lieferant: CUSTOM MADE CHEMICALS LAB
CAS-Nummer:
Spezifikationen
- Matrix:5% HNO₃
- Pk:500 mL
- Description:Wavelength calibration solution, concentrate, for ICP-OES
Spezifikationen
Über diesen Artikel
500 ml solution contains 50 mg/L Al, As, Ba,Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn and 500 mg/L K in 5% HNO₃.
Shelf life upto 36 Months, store under normal laboratory conditions, at temperatures between 15 °C to 25 °C.
Certifications: ISO 17034/17025/9001