Orders LinkContactUsLinkComponent
Screw Cap Microtubes, with Cap
Screw Cap Microtubes, with Cap
Supplier:   DELTALAB
Screw Cap Microtubes, with Cap
Supplier:  DELTALAB
Certificates
Screw Cap Microtubes, with Cap
3 Options Available Below
Product Details & Documents
Screw cap microtubes, with cap in medical grade, transparent polypropylene. The cap embodies a non-reactive ethylene-propylene o-ring. Suitable for autoclave, liquid nitrogen (gas) and boiling processes. Perfect for long term storage.
  • Withstand temperatures from –190 ℃
  • Centrifugation resistance: 20000×g
  • DNAse, RNAse, DNA, and PCR inhibitors free
  • Graduated models feature a frosted area for writing
  • The non-graduated model incorporates an external grip for an easy handling.
Packaging: Microtubes are supplied capped, in bags of 50 units.
Product Family Options
Product Information
  • Capacity
    Colour
    Material
    Sterile
    Availability
    Price
  • DELT409115/2
    Non skirted microtube with graduation
    Non skirted microtube with graduation
    1,5 ml
    Transparent
    PP
    +
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 1 950,00 €
    1 950,00 €
    1 * 2.500 ST
       1 950,00 €
  • DELT409115/3
    Non skirted microtube with graduation
    Non skirted microtube with graduation
    2 ml
    Transparent
    PP
    +
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 1 890,00 €
    1 890,00 €
    1 * 2.500 ST
       1 890,00 €
  • DELT409115/6
    Skirted microtube with graduation
    Skirted microtube with graduation
    2 ml
    Transparent
    PP
    +
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 1 850,00 €
    1 850,00 €
    1 * 2.500 ST
       1 850,00 €