ContactUsLinkComponent
CoreShield™ Microfoam Nitrile Coating Gloves
CoreShield™ Microfoam Nitrile Coating Gloves
Catalog # DALL21-1515B/10XL
Supplier:  HONEYWELL SAFETY
CoreShield™ Microfoam Nitrile Coating Gloves
Catalog # DALL21-1515B/10XL
Supplier:  HONEYWELL SAFETY
Specifications
  • Glove size:
    XL
  • Ambidextrous / hand-specific:
    Hand-specific
  • Disposable / reusable:
    Reusable
  • Colour:
    Black
  • EN norm:
    EN ISO 388:2016 (4X31A)
  • Cuff style:
    Knit wrist
  • Liner material:
    Nylon
  • Gauge:
    15
  • Finish:
    Nitrile micro-foam black coating
  • Pk:
    10
Specifications
About this item
All in one glove with enhanced cut resistance, comfort, ease of selection, cost effectiveness. Due to its high-performance fiber, the Honeywell CoreShield™ range of 22 styles offers complete cut protection from level A/A1 to F/A9, in a choice of four various gauges (from 10 gg to 18 gg) and three different coatings.
  • Easy to wear
  • Thumb reinforcement
  • Touchscreen capability for resistive screen machines
  • Silicone-free
  • Hand and wrist protection
CoreShield™ High Performance micro-foam nitrile coating provides 360° breathability, excellent grip performance, water and oil proof coating, abrasion resistance, reduced hand fatigue and excellent dexterity.
CoreShield™ Engineered Yarn: Skin friendly and comfortable material to shield the cut resistant material. Softer skin-friendly yarn in contact with the skin.
Industries: Manufacturing, automotive, metal stamping, glass handling, construction, logistics.
Certifications: EN ISO 388:2016 (4X31A), ANSI/ISEA 105 (A1).
Packaging: Gloves are packed in a plastic bag by inner pack of 10 pairs, 10 inner packs per case (100 pairs).