Anti-PIP2;1, PIP2;2, PIP2;3 Rabbit Polyclonal Antibody
AGRIAS224810
: Agrisera
- Antibody type:Primary
- Antigen name:Plasma membrane intrinistic protein 2-1, 2-2, 2-3
- Antigen symbol:PIP2;1, PIP2;2, PIP2;3
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Arabidopsis
- Format:Lyophilised
- Antigen synonyms:Aquaporin PIP2-3|plasma membrane intrinsic protein 2c|PIP2b|plasma membrane intrinsic protein 2a|TMP2C|PIP2a|RD28-PIP|aquaporin PIP2-2|TMP2b|aquaporin PIP2-1|plasma membrane intrinsic protein 2b|(WSII-TIP)|water stress-induced tonoplast intrinsic protein|PIP2c
- Purity:Affinity purified serum, in PBS pH 7,4
- Storage temperature:Store lyophilised/reconstituted at −20 °C, once reconstituted make aliquots to avoid repeated freeze-thaw cycles.
- Immunogen:KLH-conjugated synthetic peptide derived from Arabidopsis thaliana PIP2 proteins: P43286, At3g53420 , AtPIP2-2 P43287, At2g37170 , AtPIP2-3 P30302, At2g37180
- Tested applications:Western blot (WB)
- Pk:50 µG
PIP2;2 is a plasma membrane aquaporin.
- For reconstitution add 50 µl, of sterile or deionised water
Spin tubes briefly prior to opening them to avoid any losses that might occur from lyophilised material adhering to the cap or sides of the tubes.
Recommended dilution: 1:1000 (WB).
Molecular weight: 3,4 kDa (due to N-terminal or C-terminal processing).
: For research use only.
Type: Primary
Antigen: PIP2;1, PIP2;2, PIP2;3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Arabidopsis thaliana