Order Entry
Austria
ContactUsLinkComponent
 
L-Alanine
Catalog # A1688.0100
Supplier:  PanReac AppliChem
CAS Number:  
L-Alanine
Catalog # A1688.0100
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :89.09 g/mol
Merck Index:13,00203
Boiling Pt:189 °C (1013 hPa)
Melting Pt:79 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-273-8
CAS Number:56-41-7
Formula:C₃H₇NO₂
MDL Number:MFCD00064410