ContactUsLinkComponent
Cellcyte Gen 2 Live Cell Imaging and Analysis
Cellcyte Gen 2 Live Cell Imaging and Analysis
Supplier:  DISCOVER ECHO
Certificates
undefined
Cellcyte Gen 2 Live Cell Imaging and Analysis
Supplier:  DISCOVER ECHO
Specifications
  • Catalog No:
  • 630-3558
  • 630-3561
  • Observation method:
  • Fluorescence and phase contrast
  • Magnification:
  • 4×, 10×, 20×
  • Camera:
  • 5 MP sCMOS monochrome camera (2448×2048 pixels)
  • Sensor:
  • CMOS
  • Display:
  • 55,88 cm (22") HD monitor
  • Output:
  • USB
  • Electrical:
  • 100 - 240 V (50/60 Hz)
Specifications
Related Information
Objective lenses
Product Information