ContactUsLinkComponent
Manual Vacuum Regulation Turret with On and Off, Flush Mount
Manual Vacuum Regulation Turret with On and Off, Flush Mount
Catalog # ILMV700563
Manual Vacuum Regulation Turret with On and Off, Flush Mount
Catalog # ILMV700563
Specifications
  • Description:
    On/Off with manual flow control turrets, flush mount, 2 screw mount, for use with wood furniture
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Quick opening/closing via ball valve of vacuum line plus manual vacuum control.
  • Dimensions: 690 mm
  • Diameter: 820 mm protrusion from wall and 20 mm into wall
Stainless steel On/Off ball valve allows easy repeat of flow rate setting.
Connect PTFE 10 mm ext.Ø tubing to valve using a PVDF compression fitting. SS Ball Valve has stepped hose barb that accepts 6,35 to 9,52 mm vacuum hose (DN 8-10).