ContactUsLinkComponent
Self-Laminating Aggressive Adhesive Vinyl Wrap Around Labels for Wraptor A6200 Printers
Self-Laminating Aggressive Adhesive Vinyl Wrap Around Labels for Wraptor A6200 Printers
Catalog # BRDY152201
Supplier:  Brady
Self-Laminating Aggressive Adhesive Vinyl Wrap Around Labels for Wraptor A6200 Printers
Catalog # BRDY152201
Supplier:  Brady
Specifications
  • Colour:
    White/clear
  • Description:
    Compatible for Wraptor A6200 printers
  • Dimensions:
    12,70×31,75 mm
  • Material:
    Self-laminating Vinyl
  • Packed:
    Roll of 450 labels
  • Pk:
    1 Roll
  • Adhesive:
    Permanent acrylic
Specifications
About this item
Keep your wire identification secure with this self-laminating marker with extra-strong adhesive.
  • The extra-strong adhesive makes these markers ideal for small-gauge wires
  • Features a white printable area and a clear tail that wraps around the marker
  • Tail serves as an overlaminate to protect the print
  • A ribbon is required to print this material
Abrasion-Resistant, chemical-resistant, humidity-resistant, oil-resistant, water-resistant, weather-resistant.
Certifications: Compliant with Directive 2011/65/EU and 2015/863.