ISO Safety Signs - Warning; Substance or Mixture that can Cause an Environmental Hazard
Artikelnr. BRDY309879
Lieferant: Brady
Spezifikationen
- Description:ISO Safety sign - Warning; Substance or mixture that can cause an environmental hazard, 7 signs per pack
- Dimensions:50×43 mm
- Material:PE
- Pk:1
Spezifikationen
Über diesen Artikel
ISO safety sign depicts 'Warning; Substance or Mixture that can Cause an Environmental Hazard' message. It helps to avoid any possibility to have contact with or spills of substances or mixtures considered hazardous to health.
- Triangle shape
- Black on yellow finish
- 0,099 mm thickness
- Permanent acrylic adhesive and glossy finish
Applications includes facility identification, hazard communication identification.
Certifications: Compliant with Directive 2011/65/EU and 2015/863.