Order Entry
Austria
ContactUsLinkComponent
Supelco® BIOshell™ IgG 1000 Å C18 Fused-Core® HPLC Columns
Supelco® BIOshell™ IgG 1000 Å C18 Fused-Core® HPLC Columns
Supplier:  Merck
Certificates
Supelco® BIOshell™ IgG 1000 Å C18 Fused-Core® HPLC Columns
Supplier:  Merck

Specifications

  • Catalog No:
  • 581349-U
  • 581360-U
  • 581362-U
  • 581348-U
  • 581382-U
  • 582701-U
  • 582710-U
  • 582713-U
  • 581361-U
  • 582708-U
  • 582709-U
  • 582706-U
  • 582707-U
  • 582704-U
  • 581367-U
  • 582705-U
  • 581366-U
  • 582703-U
  • 582715-U
  • 581383-U
  • 581381-U
  • 581380-U
  • Description:
  • HPLC guard cartridges, pack of 3
  • HPLC guard cartridges, pack of 3
  • HPLC column
  • HPLC column
  • HPLC column
  • HPLC column
  • HPLC column
  • HPLC column
  • HPLC guard cartridges, pack of 3
  • HPLC column
  • HPLC column
  • HPLC column
  • HPLC column
  • HPLC column
  • HPLC column
  • HPLC column
  • HPLC column
  • HPLC column
  • HPLC column
  • HPLC column
  • HPLC column
  • HPLC column
  • Application:
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • For separating high molecular weight compounds such as proteins
    mAbs
    ADCs
    Biosimilars
    H/D Exchange
    mAb Fragments
  • Carbon load:
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • 1,40%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Max. pressure:
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • pH range:
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • Surface area:
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • 22 m²/g
  • Temperature limit:
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • USP code:
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1

Specifications

Frequently Bought Together