Order Entry
Austria
ContactUsLinkComponent
Hydrofluoric acid 48% ACS
Hydrofluoric acid 48% ACS
Catalog # SIAL695068-25ML
Supplier:  Merck
CAS Number:  
Hydrofluoric acid 48% ACS
Catalog # SIAL695068-25ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    25 mL
  • Seq:
    2315
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    48%

Specification Test Results

About this item

 :
 :20.01 g/mol
EINECS:231-634-8
UN:1790
 :GHS06
 :GHS05
Storage Temperature:Ambient
CAS Number:7664-39-3
Formula:HF
MDL Number:MFCD00011346
ADR:8 II