Order Entry
Austria
ContactUsLinkComponent
Supelco® BIOshell™ HPLC Columns, 160 Å Peptide CN Fused-Core®
Supelco® BIOshell™ HPLC Columns, 160 Å Peptide CN Fused-Core®
Supplier:  Merck
Certificates
undefined
Supelco® BIOshell™ HPLC Columns, 160 Å Peptide CN Fused-Core®
Supplier:  Merck

Specifications

  • Catalog No:
  • 67068-U
  • 66969-U
  • 66968-U
  • 67069-U
  • 66970-U
  • 67066-U
  • 66966-U
  • 67067-U
  • 66967-U
  • 66973-U
  • 67071-U
  • 66971-U
  • 67080-U
  • 66972-U
  • 67064-U
  • 67065-U
  • 66965-U
  • 67062-U
  • 67063-U
  • 66978-U
  • 67165-U
  • 66979-U
  • 67061-U
  • 67081-U
  • 66976-U
  • 67161-U
  • 67084-U
  • 67163-U
  • 66977-U
  • 67164-U
  • 67082-U
  • 67159-U
  • 66974-U
  • 67160-U
  • 67083-U
  • 66975-U
  • Application:
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Carbon load:
  • 1,5%
  • 2,2%
  • 2,2%
  • 1,5%
  • 2,2%
  • 1,5%
  • 2,2%
  • 1,5%
  • 2,2%
  • 2,2%
  • 1,5%
  • 2,2%
  • 1,5%
  • 2,2%
  • 1,5%
  • 1,5%
  • 2,2%
  • 1,5%
  • 1,5%
  • 2,2%
  • 2,2%
  • 2,2%
  • 1,5%
  • 1,5%
  • 2,2%
  • 2,2%
  • 1,5%
  • 2,2%
  • 2,2%
  • 2,2%
  • 1,5%
  • 2,2%
  • 2,2%
  • 2,2%
  • 1,5%
  • 2,2%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Max. pressure:
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • pH range:
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • 1 - 8
  • Surface area:
  • 60 m²/g
  • 90 m²/g
  • 90 m²/g
  • 60 m²/g
  • 90 m²/g
  • 60 m²/g
  • 90 m²/g
  • 60 m²/g
  • 90 m²/g
  • 90 m²/g
  • 60 m²/g
  • 90 m²/g
  • 60 m²/g
  • 90 m²/g
  • 60 m²/g
  • 60 m²/g
  • 90 m²/g
  • 60 m²/g
  • 60 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 60 m²/g
  • 60 m²/g
  • 90 m²/g
  • 90 m²/g
  • 60 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 60 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 60 m²/g
  • 90 m²/g
  • Temperature limit:
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • USP code:
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10
  • L10

Specifications

Related Information

Guard Cartridge Holder

Product Information