ContactUsLinkComponent
Supelco® BIOshell™ Guard Cartridge Holder
Catalog # 66841-U
Supplier:  Merck
Supelco® BIOshell™ Guard Cartridge Holder
Catalog # 66841-U
Supplier:  Merck
Specifications
  • Description:
    Guard cartridge holder
  • Pk:
    1
Specifications
About this item
This guard cartridge holder can be used with 4,6/3,0 and 2,1 mm Int.Ø BIOShell™ columns.