Order Entry
Austria
ContactUsLinkComponent
Thallium Standard Solution, 1000 mg/L Tl in Diluted Nitric Acid for ICP-MS
Thallium Standard Solution, 1000 mg/L Tl in Diluted Nitric Acid for ICP-MS
Catalog # ACSDM262.2NP.L1
Supplier:  CPACHEM
CAS Number:  
Thallium Standard Solution, 1000 mg/L Tl in Diluted Nitric Acid for ICP-MS
Catalog # ACSDM262.2NP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Thallium
  • Symbol:
    Tl

Specifications

About this item