Order Entry
Austria
ContactUsLinkComponent
Titanium Standard Solution, 1000 mg/L Ti in Diluted Nitric Acid for ICP-MS
Titanium Standard Solution, 1000 mg/L Ti in Diluted Nitric Acid for ICP-MS
Catalog # ACSDM261.2N2FP.L1
Supplier:  CPACHEM
CAS Number:  
Titanium Standard Solution, 1000 mg/L Ti in Diluted Nitric Acid for ICP-MS
Catalog # ACSDM261.2N2FP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃/2% HF
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Titanium
  • Symbol:
    Ti

Specifications

About this item